missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuroligin 2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518668
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
Specifications
| Neuroligin 2 | |
| Polyclonal | |
| Unconjugated | |
| NLGN2 | |
| Kiaa1366; neuroligin 2; neuroligin-2; NL2; Nlgn2; ortholog of human and rat neuroligin 2 NLGN2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 57555 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8NFZ4 | |
| NLGN2 | |
| Synthetic peptide sequence (NPPDTDIRDPGKK) corresponding to the internal amino acids of NLGN2 (aa 165-178). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction