missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 (CD304) Mouse anti-Human, PE, Clone: U21-1283, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565952
Additional Details : Weight : 0.01000kg
Description
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.Specifications
Neuropilin-1 (CD304) | |
Monoclonal | |
PE | |
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R | |
Recombinant Human Neuropilin-1 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
IgG2b κ |
Flow Cytometry | |
U21-1283 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Cell Biology | |
Human | |
Purified |
For Research Use Only.