missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 Recombinant Rabbit Monoclonal Antibody (ARC0488), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA535253
This item is not returnable.
View return policy
Description
Immunogen sequence: IDETGSTPGY EGEGEGDKNI SRKPGNVLKT LDPILITIIA MSALGVLLGA VCGVVLYCAC WHNGMSERNL SALENYNFEL VDGVKLKKDK LNTQSTYSEA
Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.Specifications
Neuropilin-1 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14786, P97333, Q9QWJ9 | |
NRP1 | |
A synthetic peptide corresponding to a sequence within amino acids 824-923 of human Neuropilin-1 (NRP1) (O14786) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
ARC0488 | |
Unconjugated | |
NRP1 | |
A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18186, 246331, 8829 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |