Learn More
Contec™ NeutraKlean Detergent
Ideal for the low level cleaning required after maintenance and to remove disinfectant residues and general soil.
Brand: Contec™ FBC503NK
75.70 GBP valid until 2024-05-31
Use promo code "21724" to get your promotional price.
Additional Details : CAS Number : 2605-79-0 Weight : 9.77400kg
Description
- Free from contamination and particulates, filtered to 0.22μm under Grade A airflow in Grade C cleanroom
- Low foaming detergent action
- Low level of surfactant
- Double bagged packed in linear tear packaging
Specifications
NeutraKlean Detergent | |
0.3% amine oxide in purified water (EP) | |
For cleaning of life science cleanrooms |
Non-sterile | |
0.22μm | |
5 L |