Learn More
Neutral Glass Specimen Tubes
Use these tubes for a variety of applications. Neutral Glass Specimen Tubes are ideal for samples which are susceptible to pH change.
£9.73 - £606.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16348665
|
Samco
T103/V3 |
14 mL |
£63.65
Pack of 162 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
13291630
|
Samco
T103/V3 |
14 mL |
£9.73
Pack of 162 |
In Stock
Log in to see stock. |
|||||
13130523
|
Samco
T101/V1 |
1.75 mL |
£606.00
Pack of 882 |
In Stock
Log in to see stock. |
|||||
Description
- Tall specimen tubes
- Neutral glass with screw neck
- Good chemical resistance
- Should not be taken from cold and placed directly into the hottest part of the flame without pre-heating
Specifications
Neutral Glass |