missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutral Glass Test Tubes
Use these test tubes for general mixing and simple laboratory work. Neutral Glass Test Tubes will withstand moderate warming (ex. in a water bath).
Brand: Samco G022/33
Code : 02
Additional Details : Weight : 0.77940kg
Description
- Medium walled test tubes with rim
- Should not be placed directly into naked flames
- Available in packs of 100
Specifications
ISO 4142:2002 | |
Samples which are susceptible to pH change | |
12 mm |
100 mm | |
Rim |