Learn More
Thermo Scientific Chemicals Neutral Red, pure, high purity biological stain
£80.90 - £225.00
Chemical Identifiers
CAS | 553-24-2 |
---|---|
Molecular Formula | C15H17ClN4 |
Molecular Weight (g/mol) | 288.779 |
MDL Number | MFCD00012651 |
InChI Key | PGSADBUBUOPOJS-UHFFFAOYSA-N |
Synonym | Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride |
PubChem CID | 11105 |
ChEBI | CHEBI:86370 |
IUPAC Name | 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride |
SMILES | CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10420891
|
Thermo Scientific Chemicals
229810250 |
25 g | Glass bottle |
£80.90
25g |
In Stock
Log in to see stock. |
||||
10423265
|
Thermo Scientific Chemicals
229811000 |
100 g | Glass bottle |
£225.00
100g |
Estimated Shipment: 17-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
553-24-2 | |
288.779 | |
PGSADBUBUOPOJS-UHFFFAOYSA-N | |
11105 | |
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride |
C15H17ClN4 | |
MFCD00012651 | |
Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride | |
CHEBI:86370 | |
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl |
Specifications
290.0°C | |
50% min. | |
MFCD00012651 | |
15, 6573 | |
Solubility in water: 50g/L. Other solubilities: 1.8% in abs. alcohol, 3.75% in cellosolve, 3.0% in ethylene glycol, practically insoluble in xylene | |
Authentic | |
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride | |
11105 | |
288.77 | |
Green | |
Neutral red, High purity biological stain |
553-24-2 | |
C15H17ClN4 | |
25, 401 | |
Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride | |
PGSADBUBUOPOJS-UHFFFAOYSA-N | |
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl | |
288.779 | |
CHEBI:86370 | |
Pure | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
GHS P Statement:
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only