missing translation for 'onlineSavingsMsg'
Learn More

Neutral Red, Pure, Indicator Grade, C.I.50040, Fisher Chemical

Product Code. 10080290
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Melting Point 290°C
CAS 553-24-2
Molecular Formula C15H17ClN4
MDL Number 12651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
Molecular Weight (g/mol) 288.779
PubChem CID 11105
ChEBI CHEBI:86370
Formula Weight 288.77g/mol
Packaging Glass Jar
Color Green
Quantity 5 g
Physical Form Powder
Show More Show Less
Product Identifier
  • Neutral Red
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'