missing translation for 'onlineSavingsMsg'
Learn More

Nevirapine, 98%, Thermo Scientific Chemicals

£57.79 - £173.00

Chemical Identifiers

CAS 129618-40-2
Molecular Formula C15H14N4O
Molecular Weight (g/mol) 266.3
InChI Key NQDJXKOVJZTUJA-UHFFFAOYSA-N
Synonym nevirapine, viramune, bi-rg-587, viramune xr, nvp, birg587, 11-cyclopropyl-4-methyl-5,11-dihydro-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one, nevirapine usan:inn, unii-99dk7fvk1h, 11-cyclopropyl-5,11-dihydro-4-methyl-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one
PubChem CID 4463
ChEBI CHEBI:63613
IUPAC Name 11-cyclopropyl-4-methyl-5H-dipyrido[2,3-e
SMILES CC1=C2C(=NC=C1)N(C3=C(C=CC=N3)C(=O)N2)C4CC4
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15517729
View Documents
Thermo Scientific Acros
461152500
250 mg
£57.79
250mg
Please to purchase this item. Need a web account? Register with us today!
15507729
View Documents
Thermo Scientific Acros
461150010
1 g
£173.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

129618-40-2
266.3
nevirapine, viramune, bi-rg-587, viramune xr, nvp, birg587, 11-cyclopropyl-4-methyl-5,11-dihydro-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one, nevirapine usan:inn, unii-99dk7fvk1h, 11-cyclopropyl-5,11-dihydro-4-methyl-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one
CHEBI:63613
CC1=C2C(=NC=C1)N(C3=C(C=CC=N3)C(=O)N2)C4CC4
C15H14N4O
NQDJXKOVJZTUJA-UHFFFAOYSA-N
4463
11-cyclopropyl-4-methyl-5H-dipyrido[2,3-e

Specifications

129618-40-2
Authentic
C15H14N4O
NQDJXKOVJZTUJA-UHFFFAOYSA-N
11-cyclopropyl-4-methyl-5H-dipyrido[2,3-e
4463
266.3
Nevirapine
247.0°C to 249.0°C
97.5% min. (HPLC)
nevirapine, viramune, bi-rg-587, viramune xr, nvp, birg587, 11-cyclopropyl-4-methyl-5,11-dihydro-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one, nevirapine usan:inn, unii-99dk7fvk1h, 11-cyclopropyl-5,11-dihydro-4-methyl-6h-dipyrido 3,2-b:2',3'-e 1,4 diazepin-6-one
CC1=C2C(=NC=C1)N(C3=C(C=CC=N3)C(=O)N2)C4CC4
266.3
CHEBI:63613
98%
Videos
Safety and Handling

Safety and Handling

Harmful if swallowed, Harmful to aquatic life with long lasting effects

Acute toxicity (category 4), Hazardous to the aquatic environment (chronic category 3)

Product Identifier
  • Nevirapine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only