missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Scientific Test Tube Racks

Adjustable angle and tube height

Brand:  Eppendorf™ M1289-0004

343.00 GBP valid until 2024-12-31
Use promo code "23275" to get your promotional price.



Code : 99

Additional Details : Weight : 2.94830kg

 View more versions of this product

Product Code. 11319934

  • £361.00 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Inserts to help retain tubes and prevent them from spinning in the rack
  • Finger-tight knobs to quickly adjust angle, or release the rack from the base plate, for easy transport to the bench
  • Stainless-steel

Specifications

Specifications

Test Tube Rack
18 to 21 mm
Two adjustable bars and non-skid rubber mat
Stainless Steel
Biological shaker
Small
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services