missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Coccine, pure

£35.68 - £104.10

Chemical Identifiers

CAS 2611-82-7
Molecular Formula C20H11N2Na3O10S3
Molecular Weight (g/mol) 604.46
MDL Number MFCD00004084
InChI Key IVNZBWNBYXERPK-DZGBHZPSSA-K
Synonym Acid Red 18, C.I. 16255, Cochineal Red A
PubChem CID 131851859
IUPAC Name (8Z)-7-oxo-8-[(4-sulfonaphthalen-1-yl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
SMILES [Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=C4C=CC=CC4=C(C=C3)S([O-])(=O)=O)C(=O)C=C2)C(=C1)S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10583361
SDS View Documents
Thermo Scientific Chemicals
190230250
25 g Glass bottle
£35.68
25g
Please to purchase this item. Need a web account? Register with us today!
10055943
SDS View Documents
Thermo Scientific Chemicals
190231000
100 g Glass bottle
£104.10
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 220-036-2

missing translation for 'rtecsNumber' : QJ6530000

missing translation for 'tsca' : TSCA

Product Identifier
  • New Coccine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals New Coccine, pure

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.