missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals New Methylene Blue N, zinc chloride double salt, pure

Product Code. 10427780
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance (c=5mg/l; water), (c=5mg/l; water) at 0nm
Molecular Formula 0·5 ZnCl2
MDL Number MFCD00078135
Synonym Basic Blue 24, C.I. 52030
Infrared Spectrum Authentic
Lambda Max 628 to 634nm, Molar Absorption Coefficient 40000 min., 588 to 594nm, Molar Absorption Coefficient 36000 min., 284 to 290nm, Molar Absorption Coefficient 28000 min., 242 to 248nm, Molar Absorption Coefficient 16000 min.
Formula Weight 416.04
Grade Pure
Packaging Glass bottle
Color Brown to Purple
Quantity 10 g
Physical Form Crystalline Powder
Chemical Name or Material New Methylene Blue N, zinc chloride double salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'