missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NF-kB Activation Inhibitor VI, BOT-64

Product Code. 15416459
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor

A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113760-29-5
Molecular Formula C15H15NO2S, C15H15NO2S
Synonym (Z)-6,6-Dimethyl-2-(phenylimino)-6,7-dihydrobenzo[d][1,3]oxathiol-4(5H)-one

Specifications

Quantity 5 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor
Formula Weight 273.40g/mol
Sensitivity Light Sensitive
Solubility Soluble in ethanol or DMSO
Product Type Signal Transduction Reagent
Form Solid
InChI Key XFSSJIQCIPSBHJ-UHFFFAOYSA-N
SMILES CC1(C)CC2=C(SC(O2)=NC2=CC=CC=C2)C(=O)C1
IUPAC Name 6,6-dimethyl-2-(phenylimino)-4,5,6,7-tetrahydro-2H-1,3-benzoxathiol-4-one
Molecular Weight (g/mol) 273.35
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'