missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NF-kB Activation Inhibitor VI, BOT-64

Product Code. p-7082413
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 15416459

Brand: Thermo Scientific Chemicals J64555.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor

A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113760-29-5
Molecular Formula C15H15NO2S, C15H15NO2S
Synonym (Z)-6,6-Dimethyl-2-(phenylimino)-6,7-dihydrobenzo[d][1,3]oxathiol-4(5H)-one

Specifications

Quantity 5 mg
Description Off-white
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable benzoxathiole compound that acts as an IKK-2 inhibitor
Formula Weight 273.40g/mol
Sensitivity Light Sensitive
Solubility Soluble in ethanol or DMSO
Product Type Signal Transduction Reagent
Form Solid
InChI Key XFSSJIQCIPSBHJ-UHFFFAOYSA-N
SMILES CC1(C)CC2=C(SC(O2)=NC2=CC=CC=C2)C(=O)C1
IUPAC Name 6,6-dimethyl-2-(phenylimino)-4,5,6,7-tetrahydro-2H-1,3-benzoxathiol-4-one
Molecular Weight (g/mol) 273.35
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.