missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals NG-Monomethyl-L-arginine monoacetate, 99%
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 180°C to 190°C |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Color | White |
Assay Percent Range | 99% |
Infrared Spectrum | Authentic |
Merck Index | 15, 6096 |
Packaging | Glass Bottle |
Specific Rotation | +9° to +13° (20°C, 589nm) (c=4, water) |
Formula Weight | 248.3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'