missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHP2L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522010
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Xenopus laevis (92%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5' stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.Specifications
NHP2L1 | |
Polyclonal | |
Unconjugated | |
SNU13 | |
[U4/U6.U5] tri-snRNP 15.5 kD RNA binding protein; 15.5K; CTA-216E10.8; FA1; FA-1; Fertilization antigen 1; fertilization antigen-1; Fta1; high mobility group-like nuclear protein 2 homolog 1; hSNU13; NHP2 non-histone chromosome protein 2-like 1; NHP2 non-histone chromosome protein 2-like 1 (S. cerevisiae); NHP2L1; NHP2-like protein 1; NHP2-like protein 1, N-terminally processed; NHPX; non-histone chromosome protein 2-like 1; Otk27; small nuclear ribonucleoprotein 15.5kDa (U4/U6.U5); SNRNP15-5; SNU13; SNU13 homolog; SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5); SPAG12; sperm specific antigen 1; sperm-specific antigen 1; Ssfa1; U4/U6.U5 small nuclear ribonucleoprotein SNU13; U4/U6.U5 tri-snRNP 15.5 kDa protein; U4/U6.U5] tri-snRNP 15.5 kDa protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20826, 300092, 4809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P55769, Q9D0T1 | |
Nhp2l1, SNU13 | |
Full length human NHP2L1 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |