missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niacin, U.S.P. - F.C.C., J.T. Baker™
Click to view available options
Quantity:
25 g
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Specifications
Specifications
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 25 g |
Specific Gravity | 1L = 1.47kg |
Formula Weight | 123.11 |
Grade | FCC/USP |
Chemical Name or Material | Niacin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'