missing translation for 'onlineSavingsMsg'
Learn More

Microspec™ Nichrome Wire Loops

Used for bacteriological, microbiological applications and flame tests.

Brand:  Microspec™ S5

 View more versions of this product

Product Code. 15712175

  • £75.00 / Pack of 25
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Used for bacteriological and other microbiological applications and flame tests
  • Tightly wound
  • Easy change
  • Wire loops
  • Nichrome wire
Specifications

Specifications

Reusable
0.163 in.
3.54 to 3.94 in.
Nickel Chromium Wire
Silver
Wire Loop Handle
25
Yes
5 μL
4.15 mm
90 to 100 mm
Non-sterile
Inoculating Product
25 wire loops in a vial
Plastic Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.