missing translation for 'onlineSavingsMsg'
Learn More

Nickel acetylacetonate, 96%, Thermo Scientific Chemicals

Product Code. 10251123
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
IUPAC Name nickel(2+);(E)-4-oxopent-2-en-2-olate
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 230°C
Color Yellow
Boiling Point 220°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Complexometry)
Packaging Glass bottle
Quantity 5 g
Fieser 05,471; 06,417; 07,250; 08,42; 17,201
Merck Index 15, 6585
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,chloroform and benzene,insoluble in ether and ligroin
Formula Weight 256.9
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Nickel acetylacetonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel acetylacetonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'