missing translation for 'onlineSavingsMsg'
Learn More

Nickel Aluminum, Thermo Scientific Chemicals

Wear resistant coatings

£248.24

Chemical Identifiers

CAS 12635-27-7
Molecular Formula AlNi
Molecular Weight (g/mol) 85.675
MDL Number MFCD00167392
InChI Key NPXOKRUENSOPAO-UHFFFAOYSA-N
Synonym Aluminum Nickel
PubChem CID 6336846
IUPAC Name aluminum;nickel
SMILES [Al].[Ni]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359147
SDS View Documents
Thermo Scientific Alfa Aesar
039828.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Nickel Aluminum is used as wear resistant coatings. It is also used for the preparation of Raney™ Nickel catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel Aluminum, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.