missing translation for 'onlineSavingsMsg'
Learn More

Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals

£29.85 - £99.90

Chemical Identifiers

CAS 7785-20-8
Molecular Formula H8N2NiO8S2·6H2O
MDL Number MFCD00150260
Synonym Ammonium nickel sulfate hexahydrate
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11438367
SDS View Documents
Thermo Scientific Alfa Aesar
A18441.22
100 g
£29.85
100g
Please to purchase this item. Need a web account? Register with us today!
11448367
SDS View Documents
Thermo Scientific Alfa Aesar
A18441.36
500 g
£99.90
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nickel ammonium sulfate hexahydrate is used as a nickel electrolyte for electroplating and as an analytical reagent. It is also used as a soil fertilizer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

7785-20-8
H8N2NiO8S2·6H2O
MFCD00150260
Ammonium nickel sulfate hexahydrate

Specifications

7785-20-8
Ni(NH4)2(SO4)2·6H2O
UN3077
Ammonium nickel sulfate hexahydrate
394.99 (286.90 Anhydrous)
Odorless
Nickel ammonium sulfate hexahydrate
H8N2NiO8S2·6H2O
MFCD00150260
14,6501
Soluble in water.
98%
1.923
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel ammonium sulfate hexahydrate, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.