missing translation for 'onlineSavingsMsg'
Learn More

Nickel bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15492837
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Nickel(II) Bis(trifluoromethanesulfonyl)imide is used as Surface treatment, Synthesis and catalysis. It is also used as a ionic liquid in synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207861-63-0
Molecular Formula C4F12N2NiO8S4
Molecular Weight (g/mol) 618.964
MDL Number MFCD23380174
InChI Key VFUSIQAHVPEYFD-UHFFFAOYSA-N
Synonym nickel bis-trifluoromethanesulfonimide
PubChem CID 131875825
IUPAC Name bis(trifluoromethylsulfonyl)azanide;nickel(2+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ni+2]

Specifications

Color Green
Physical Form Solid
Quantity 1 g
Linear Formula [(CF3SO2)2N]2Ni
UN Number UN3261
Solubility Information It is soluble in water.
Formula Weight 618.96
Sensitivity Hygroscopic
Chemical Name or Material Nickel bis(trifluoromethylsulfonyl)imide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel bis(trifluoromethylsulfonyl)imide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Nickel bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'