Learn More
Nickel chromite, Thermo Scientific Chemicals
£82.71 - £234.17
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313587
|
Thermo Scientific Alfa Aesar
019424.14 |
25 g |
£82.71
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323587
|
Thermo Scientific Alfa Aesar
019424.22 |
100 g |
£234.17
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In mixing Nickel (II) Chromate with Aluminates or Cobalt Chromate, a binary compound known as a spinel is formed that is able to withstand temperatures up to 1000 °C. The products from these mixings can be used as oxidation-resistive components in airplane and space shuttle coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12018-18-7 | |
MFCD00064806 |
Specifications
12018-18-7 | |
NiCr2O4 | |
Insoluble in water. Soluble in hydrochloric acid. | |
Odorless |
Powder | |
MFCD00064806 | |
226.7 | |
Nickel chromite |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
missing translation for 'einecsNumber' : 234-636-7
missing translation for 'rtecsNumber' : GB9000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only