missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chromium lump, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11353378
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
11106-97-1
CrNi
110.69
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
11320920
chromium nickel
[Cr].[Ni]
This item is not returnable. View return policy
11106-97-1
CrNi
110.69
MFCD00673110
VNNRSPGTAMTISX-UHFFFAOYSA-N
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia
11320920
chromium nickel
[Cr].[Ni]

For vacuum deposition

For vacuum deposition. hey are also used in some dental restorations. Used as Heating Elements, Thermocouples, Wear Resistant Alloys. Used as intermediate and research. Nichrome wire is commonly used in ceramics and Pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Avg. Mol. Wt. or Mol. Wt. Range Ni:Cr; 80:20 wt%
Quantity 25 g
Solubility Information Insoluble in water.
Percent Purity 99.9%
Odor Odorless
Particle Size 3 to 12 mm
Form Sintered Lump
Assay Metals basis
Chemical Name or Material Nickel Chromium lump

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'