missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length, 99.95% (metals basis), Thermo Scientific Chemicals
£40.40 - £171.00
Chemical Identifiers
CAS | 11106-97-1 |
---|---|
Molecular Formula | CrNi |
Molecular Weight (g/mol) | 110.69 |
MDL Number | MFCD00673110 |
InChI Key | VNNRSPGTAMTISX-UHFFFAOYSA-N |
Synonym | nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia |
PubChem CID | 11320920 |
IUPAC Name | chromium nickel |
SMILES | [Cr].[Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324488
|
Thermo Scientific Alfa Aesar
042368.09 |
10 g | N/A | ||||||
11334488
|
Thermo Scientific Alfa Aesar
042368.18 |
50 g |
£171.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
11106-97-1 | |
110.69 | |
VNNRSPGTAMTISX-UHFFFAOYSA-N | |
11320920 | |
[Cr].[Ni] |
CrNi | |
MFCD00673110 | |
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
chromium nickel |
Specifications
11106-97-1 | |
CrNi | |
nickel chromium, nickel-chromium, nicr, nickel chromium lump, chromium, compd. with nickel 1:1, chromium nickel, acmc-1byoe, nickel chromium powder,-140+325 mesh, nickel chromium wire, 1.0mm 0.04in dia, nickel chromium wire, 0.25mm 0.01in dia | |
[Cr].[Ni] | |
110.69 | |
99.95% | |
≈0.22g/slug | |
(metals basis) |
Ni:Cr; 80:20 wt% | |
MFCD00673110 | |
VNNRSPGTAMTISX-UHFFFAOYSA-N | |
chromium nickel | |
11320920 | |
Odorless | |
Slug | |
Nickel Chromium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only