missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel Crucible

Nickel Electro™ Nickel Crucible is crevice free and suitable for temperatures up to 350°C.

Brand:  Nickel Electro™ 17

 View more versions of this product

Product Code. 11459589

  • £36.65 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 99.6% nickel purity
  • Crevice free
  • High thermal and electrical conductivities
  • Low gas content
  • Low vapor pressure
  • Cost effective replacement for platinum
Specifications

Specifications

Nickel
1 mm
Autoclavable
45 mm
Each
50 mL
45 mm
Crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers