Learn More
Nickel dimethylglyoxime, 99%, Thermo Scientific™
Paint pigment
£17.48 - £265.00
Chemical Identifiers
CAS | 13478-93-8 |
---|---|
Molecular Formula | C8H14N4NiO4 |
Molecular Weight (g/mol) | 288.92 |
MDL Number | MFCD00136028 |
InChI Key | UNMGLSGVXHBBPH-USBCBBGOSA-L |
Synonym | nickel dimethylglyoxime,, nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine |
PubChem CID | 15639912 |
SMILES | [Ni++].C\C(=N\O)\C(\C)=N/[O-].C\C(=N\O)\C(\C)=N/[O-] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13478-93-8 | |
288.92 | |
UNMGLSGVXHBBPH-USBCBBGOSA-L | |
15639912 |
C8H14N4NiO4 | |
MFCD00136028 | |
nickel dimethylglyoxime,, nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine | |
[Ni++].C\C(=N\O)\C(\C)=N/[O-].C\C(=N\O)\C(\C)=N/[O-] |
Specifications
13478-93-8 | |
C8H14N4NiO4 | |
nickel dimethylglyoxime,, nickel; n-z-3-nitrosobut-2-en-2-yl hydroxylamine | |
UNMGLSGVXHBBPH-USBCBBGOSA-L | |
nickel(2+) bis((Z)-[(3Z)-3-(hydroxyimino)butan-2-ylidene](oxido)amine) | |
15639912 | |
99% | |
Nickel dimethylglyoxime |
250°C (sublimation) | |
MFCD00136028 | |
Insoluble in water; Soluble in mineral acids | |
[Ni++].C\C(=N\O)\C(\C)=N/[O-].C\C(=N\O)\C(\C)=N/[O-] | |
288.92 | |
288.94 | |
Powder |
Safety and Handling
GHS H Statement
H351-H315-H319-H317-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
H315-H317-H319-H335-H350
missing translation for 'einecsNumber' : 236-782-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only