missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Nickel Electro™ aluminum Dish
Aluminum Dish
Marca: Nickel Electro™ 601
Detalles adicionales : Weight : 0.15000kg
Descripción
- Laboratory dish, aluminum, bright finish
- 55 mm Diameter x 10 mm high aluminum dish
- Supplied in standard packs of 5
Especificaciones
Autoclavable | |
Aluminum | |
Non-sterile | |
Dish | |
55 mm |
Round | |
Round | |
Tissue Culture | |
10 mm |