missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10741881

Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10741881
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10741881

Brand: Thermo Scientific Acros 223145000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-89-9
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrateShow MoreShow Less
PubChem CID 62601
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 250.0°C
Color White
Physical Form Crystalline Powder, Crystals or Flakes
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula (CH3CO2)2Ni·4H2O
Fieser 14,288; 16,308
Merck Index 14,6498
Solubility Information Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol
IUPAC Name nickel(2+);diacetate;tetrahydrate
Formula Weight 248.86
Percent Purity ≥99%
Grade Analytical
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material Nickel(II) acetate tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467147 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number223141000,223145000,223140025
Lot NumberA0467148 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number223141000,223145000,223140025
Lot NumberA0466391 Certificate TypeCertificate of Analysis Date26/09/2024 Catalog Number223141000,223145000,223140025
Lot NumberA0466237 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number223141000,223145000,223140025
Lot NumberA0466004 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number223141000,223145000,223140025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.