missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) acetate tetrahydrate, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10136990

Nickel(II) acetate tetrahydrate, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10136990
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10136990

Brand: Thermo Scientific Acros 415545000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-89-9, 373-02-4
Molecular Formula C4H14NiO8
Molecular Weight (g/mol) 248.84
MDL Number MFCD00066973
InChI Key OINIXPNQKAZCRL-UHFFFAOYSA-L
Synonym nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate
PubChem CID 62601
IUPAC Name nickel(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 250.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min
Packaging Plastic bottle
Linear Formula (CH3CO2)2Ni·4H2O
Quantity 500 g
Fieser 14,288; 16,308
Merck Index 14, 6498
Solubility Information Solubility in water: 182g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 248.86
Percent Purity 99%
Grade Extra Pure
Physical Form Powder
Chemical Name or Material Nickel(II) acetate tetrahydrate, Extra Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464357 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number415545000,415541000,415540050
Lot NumberA0451223 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number415545000,415541000,415540050
Lot NumberA0403333 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number415545000,415541000,415540050
Lot NumberA0398334 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number415545000,415541000,415540050
Lot NumberA0393421 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number415545000,415541000,415540050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) acetate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) acetate tetrahydrate, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.