missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) bromide, anhydrous, 99%

Product Code. 11478650
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11478650

Brand: Thermo Scientific Alfa Aesar 053115.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13462-88-9 | Br2Ni | 218.501 g/mol

Nickel(II) bromide is used as a source of the bromide ion. It is an important raw material and intermediate used in synthetic chemistry. Further, it is used as a catalyst in cross-coupling reactions.It is used in the preparation of paramagnetic trans-diaquabis(N,N-diethylethylenediamine) nickel(II) bromide and diamagnetic bis(N,N-diethylethylenediamine)nickel(II) bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-88-9
Molecular Formula Br2Ni
Molecular Weight (g/mol) 218.501
MDL Number MFCD00011141
InChI Key IPLJNQFXJUCRNH-UHFFFAOYSA-L
Synonym nickel bromide, nickel ii bromide, nickel dibromide, nickelous bromide, nickel bromide nibr2, nickel 2+ bromide, unii-41sgh8a2s6, nibr2, nickel ii bromide, anhydrous
PubChem CID 278492
IUPAC Name dibromonickel
SMILES [Ni](Br)Br

Specifications

Melting Point 963°C
Physical Form Crystalline
Quantity 50 g
Linear Formula NiBr2
UN Number UN3077
Merck Index 14,6502
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 218.53
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 5.098 g/mL at 27°C
Chemical Name or Material Nickel (II) bromide, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) bromide anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Nickel bromide (NiBr2)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.