Learn More
Nickel(II) bromide hydrate, 98%, for analysis, Thermo Scientific Chemicals
£62.71 - £229.14
Chemical Identifiers
CAS | 207569-11-7, 13462-88-9 |
---|---|
Molecular Formula | Br2Ni |
Molecular Weight (g/mol) | 218.50 |
MDL Number | MFCD00149806 |
InChI Key | IPLJNQFXJUCRNH-UHFFFAOYSA-L |
Synonym | nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci |
PubChem CID | 57377118 |
SMILES | [Ni++].[Br-].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10082700
|
Thermo Scientific Acros
214871000 |
100 g | Glass bottle |
£62.71
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10398200
|
Thermo Scientific Acros
214875000 |
500 g | Glass bottle |
£229.14
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207569-11-7, 13462-88-9 | |
218.50 | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
57377118 |
Br2Ni | |
MFCD00149806 | |
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci | |
[Ni++].[Br-].[Br-] |
Specifications
963.0°C | |
Orange | |
100.0 | |
Crystalline Powder or Needles | |
Br2Ni | |
MFCD00149806 | |
10, 6340 | |
Solubility in water: soluble in water. Other solubilities: soluble in ammonium hydroxide, soluble in alcohol and ether | |
[Ni++].[Br-].[Br-] | |
218.50 | |
218.53 | |
Analytical |
207569-11-7, 13462-88-9 | |
97.5 | |
5 to 7 (5% aq. soln.) | |
97.5% min. (on dry substance) (Argentometry) | |
NiBr2·xH2O | |
04,351 | |
nickel ii bromide hydrate, dibromonickel hydrate, nickel bromide hydrate, br2ni.h2o, ksc566q3p, nickel ii bromide monohydrate, nickel bromide nibr2 ,monohydrate 9ci | |
IPLJNQFXJUCRNH-UHFFFAOYSA-L | |
dibromonickel;hydrate | |
57377118 | |
98% | |
Nickel(II) bromide hydrate, 98% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
May cause cancer by inhalation.
Suspected of causing genetic defects.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure.
M
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain speci
GHS Signal Word: Danger
- Nickel(II) bromide hydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H341-Suspected of causing genetic defects.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.