missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals Product Code.: 11353758

Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals

Product Code. 11353758
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11353758

Brand: Thermo Scientific Alfa Aesar 040242.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in Soporific and sedatives pharmaceutical Industry, Photosensitive industry, Analytical Chemistry, and also electrolyte in high energy battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-49-3
Molecular Formula Br2H6NiO3
Molecular Weight (g/mol) 272.546
MDL Number MFCD00011141
InChI Key UQPSGBZICXWIAG-UHFFFAOYSA-L
Synonym nickel ii bromide trihydrate, dibromonickel trihydrate, nickel dibromide hydrate, nickel bromide trihydrate, br2ni.3h2o, ksc497q6j, dibromonickel-water 1/3, nickel ii bromide trihydrate atShow MoreShow Less
PubChem CID 53471882
IUPAC Name dibromonickel;trihydrate
SMILES O.O.O.[Ni](Br)Br

Specifications

Melting Point 300°C (in -3H2O)
Physical Form Crystalline Granules
Quantity 50 g
UN Number UN3077
Merck Index 14,6502
Solubility Information Soluble in water, alcohol
Formula Weight 272.58 (218.53 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Nickel(II) bromide trihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP08K005 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number040242.18,040242.30
Lot NumberS20J096 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number040242.18,040242.30,040242
Lot NumberS20J097 Certificate TypeCertificate of Analysis Date28/06/2023 Catalog Number040242.18,040242.30,040242
Lot NumberQ16F039 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040242.18,040242.30
Lot NumberD13X055 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040242.18,040242.30
compliance-icons compliance-icons
Product Identifier
  • Nickel(II) bromide trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Nickel(II) bromide trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) bromide trihydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.