missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) carbonate, anhydrous, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Beige |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in dilute acids. Insoluble in cold water. |
Formula Weight | 118.72 |
Appearance | Light powder |
Chemical Name or Material | Nickel(II) carbonate, anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'