missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) carbonate, anhydrous, 98%, Thermo Scientific Chemicals Product Code.: 11318127

Nickel(II) carbonate, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11318127
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11318127

Brand: Thermo Scientific Alfa Aesar 022897.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nickel(II) carbonate is used in sulphamate baths, metal phosphating, electroplating and ceramic applications. It acts as a precursor to catalysts and an intermediate in the hydrometallurgical purification of nickel from its ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3333-67-3
Molecular Formula CNiO3
Molecular Weight (g/mol) 118.70
MDL Number MFCD00011144
InChI Key ZULUUIKRFGGGTL-UHFFFAOYSA-L
Synonym nickel carbonate, nickelous carbonate, nickel ii carbonate, nickel monocarbonate, unii-wm50squ829, carbonic acid, nickel 2+ salt 1:1, nickel 2+ carbonate nico3, nickel 2+ ion carbonate, nickel carbonate nico3
PubChem CID 18746
IUPAC Name nickel(2+);carbonate
SMILES [Ni++].[O-]C([O-])=O

Specifications

Color Beige
Physical Form Powder
Quantity 250 g
Assay Percent Range 98%
Solubility Information Soluble in dilute acids. Insoluble in cold water.
Formula Weight 118.72
Appearance Light powder
Chemical Name or Material Nickel(II) carbonate, anhydrous

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM20L024 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog Number022897.30,022897.18
Lot NumberM17K010 Certificate TypeCertificate of Analysis Date23/01/2024 Catalog Number022897.30,022897.18
Lot NumberX03J003 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number022897.30,022897.18
Lot NumberX02J014 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number022897.30,022897.18
Lot NumberP28I012 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number022897.30,022897.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) carbonate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Nickel(II) carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) carbonate, anhydrous, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.