missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate (basic) hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific™

Product Code. 11316937
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155775-31-8
Molecular Formula CH4Ni3O7
Molecular Weight (g/mol) 304.12
MDL Number MFCD00016262
InChI Key ATYNVYNPWGEMMI-UHFFFAOYSA-H
Synonym nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis
PubChem CID 86176052
IUPAC Name nickel(3+);carbonate;hydroxide;hydrate
SMILES [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O

Specifications

Color Green
Physical Form Powder
Quantity 5 g
Linear Formula NiCO3·2Ni(OH)2·xH2O
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 304.18 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Nickel(II) carbonate (basic) hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'