missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni), Thermo Scientific Chemicals

Product Code. 10360644
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39430-27-8, 65405-96-1
Molecular Formula CH4Ni3O7·xH2O
Molecular Weight (g/mol) 304.11
MDL Number MFCD00150262
Synonym Basic nickel carbonate

Specifications

Color Green
Physical Form Powder
Quantity 1 kg
Linear Formula 2Ni(OH)2·xH2O
Merck Index 13, 6527
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia and in diluted acids
Formula Weight 304.11
Percent Purity ≥45% (NI)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Nickel(II) carbonate hydroxide hydrate, ca 45-47% Ni, For analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'