Learn More
Nickel(II) chloride, 98%, Thermo Scientific Chemicals
Nickel(II) chloride, 98%, Cl2Ni, CAS Number-7718-54-9, 250g, 97.5% min. (Argentometry), 100.0, CHEBI:34887, Brown to Orange, 231-743-, 129.6, GHS Signal Word: Danger
£50.25 - £174.87
Chemical Identifiers
CAS | 7718-54-9 |
---|---|
MDL Number | MFCD00011142 |
InChI Key | QMMRZOWCJAIUJA-UHFFFAOYSA-L |
PubChem CID | 24385 |
ChEBI | CHEBI:34887 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10414905
|
Thermo Scientific Acros
387320500 |
50 g | Glass bottle |
£50.25
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10222463
|
Thermo Scientific Acros
387322500 |
250 g | Glass bottle |
£174.87
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1001.0°C | |
Brown to Orange | |
100.0 | |
97.5% min. (Argentometry) | |
NiCl2 | |
15, 6590 | |
QMMRZOWCJAIUJA-UHFFFAOYSA-L | |
dichloronickel | |
24385 | |
129.6 | |
Nickel(II) chloride |
7718-54-9 | |
97.5 | |
Powder | |
Cl2Ni | |
MFCD00011142 | |
Solubility in water: slightly soluble | |
Cl[Ni]Cl | |
129.6 | |
CHEBI:34887 | |
98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-743-
- Nickel(II) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only