missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11315747
Click to view available options
Quantity:
5 g
25 g

Nickel(II) chloride is used in electroplating, electroforming ,electroless plating, tea and coffee plantation, dye mordant. It is also used in semiconducting metal-containing polymer synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Melting Point 1,001°C
Color Yellow
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula NiCl2
UN Number UN3288
Merck Index 14,6505
Solubility Information It is soluble in water,ethylene glycol,ethanol, ammonium hydroxide. It is insoluble in ammonia, nitric acid.
Formula Weight 129.6
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.55 g/mL
Assay (metals basis)
Chemical Name or Material Nickel (II) chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'