missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 99.3% (metals basis), Thermo Scientific Chemicals

Catalog Number 11381759
Click to view available options
Quantity:
500 g
2 kg
7791-20-0
Cl2Ni·6H2O
MFCD00149809
7791-20-0
Cl2Ni·6H2O
MFCD00149809

Zirconium dichloride oxide is utilized as a precursor to prepare other zirconium compounds. It is employed in acid dyes, pigment toners and antiperspirants. It finds application as a rubber additive and a fiber treatment agent. It is also used in paint drying, refractories, ceramics and glaze.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY
Melting Point 140°C-H2O
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.3% (metals basis)
Solubility Information Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide.
Formula Weight 237.71 (129.60anhydrous)
Odor Odorless
Appearance Green crystals
Density 1.92 g/cm3 at 20°C
Chemical Name or Material Nickel(II) chloride hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'