missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Melting Point | 140°C-H2O |
Color | Green |
Physical Form | Crystalline |
Quantity | 50 g |
Assay Percent Range | 99.95% (metals basis) |
Solubility Information | Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide. |
Formula Weight | 237.71 (129.60 Anhydrous) |
Odor | Odorless |
Density | 1.92 g/cm3 at 20°C |
Chemical Name or Material | Nickel(II) chloride hexahydrate |
Safety and Handling
Product Identifier
- Nickel(II) chloride hexahydrate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Nickel(II) chloridehexahydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction