missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

£37.19 - £312.56

Chemical Identifiers

CAS 69098-15-3
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.59
MDL Number MFCD00149808
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
Synonym nickel ii chloride hydrate, nickel chloride hydrate, cl2ni.h2o, nicl2.h2o, nickel 11 chloride hydrate, nickel ii chloride hydrate, puratronic, nickel ii chloride hydrate trace metals basis
PubChem CID 16212333
SMILES [Cl-].[Cl-].[Ni++]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11356937
View Documents
Thermo Scientific Alfa Aesar
010815.06
5 g
£37.19
5g
Please to purchase this item. Need a web account? Register with us today!
11346937
View Documents
Thermo Scientific Alfa Aesar
010815.14
25 g
£90.55
25g
Please to purchase this item. Need a web account? Register with us today!
11336937
View Documents
Thermo Scientific Alfa Aesar
010815.22
100 g
£312.56
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in electroplating and as a catalyst for organic conversions, for example in chemo-selective thioacetalization of aldehydes. In combination with lithium aluminum hydride, it serves as a reducing agent for alkenes, alkynes, and organic halides; it can cleave N-O bond and open epoxides. It is a precursor to several nickel-phosphine complexes, such as bis(triphenylphosphine)nickel(II) chloride, which are used in alkyne trimerizations, carbonylations, and as catalysts in organic reactions such as Suzuki-Miyaura cross coupling reactions as an alternative to palladium(0) catalysts. It is the precursor to acetylacetonate complex of Ni, used for producing 1,5-cyclooctadiene complex, an important reagent in organonickel chemistry. It can be used to prepare the sandwich compound nickelocene through dimethoxyethane complex of nickel chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69098-15-3
129.59
QMMRZOWCJAIUJA-UHFFFAOYSA-L
16212333
Cl2Ni
MFCD00149808
nickel ii chloride hydrate, nickel chloride hydrate, cl2ni.h2o, nicl2.h2o, nickel 11 chloride hydrate, nickel ii chloride hydrate, puratronic, nickel ii chloride hydrate trace metals basis
[Cl-].[Cl-].[Ni++]

Specifications

69098-15-3
Crystalline Aggregates
NiCl2·xH2O
UN3288
nickel ii chloride hydrate, nickel chloride hydrate, cl2ni.h2o, nicl2.h2o, nickel 11 chloride hydrate, nickel ii chloride hydrate, puratronic, nickel ii chloride hydrate trace metals basis
QMMRZOWCJAIUJA-UHFFFAOYSA-L
dichloronickel;hydrate
16212333
99.995%
Hygroscopic
Nickel(II) chloride hydrate
Green
Cl2Ni
MFCD00149808
14,6505
Freely soluble in water. Soluble in ethylene glycol,ethanol,and ammonium hydroxide.
[Cl-].[Cl-].[Ni++]
129.59
129.60 (Anhydrous)
Puratronic™
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H334-H350-H360-H372-H341-H315-H317
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c

H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-743-0

missing translation for 'rtecsNumber' : QR6480000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt