missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11368557

Nickel(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368557
25 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11368557

Brand: Thermo Scientific Alfa Aesar 035713.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7718-54-9
Molecular Formula Cl2Ni
Molecular Weight (g/mol) 129.593
MDL Number MFCD00011142
InChI Key QMMRZOWCJAIUJA-UHFFFAOYSA-L
PubChem CID 24385
ChEBI CHEBI:34887
IUPAC Name dichloronickel
SMILES Cl[Ni]Cl

Specifications

Melting Point 1001°C
Color Yellow to Orange
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water.
Formula Weight 129.6
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow to orange powder
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Density 3.55
Chemical Name or Material Nickel(II) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) chloride, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.