Learn More
Nickel(II) hydroxide, for analysis, Thermo Scientific Chemicals
£18.50 - £128.64
Chemical Identifiers
CAS | 12054-48-7 |
---|---|
Molecular Formula | H2NiO2 |
Molecular Weight (g/mol) | 92.71 |
MDL Number | MFCD00011140 |
InChI Key | BFDHFSHZJLFAMC-UHFFFAOYSA-L |
IUPAC Name | nickel(2+) dihydroxide |
SMILES | [OH-].[OH-].[Ni++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10155160
|
Thermo Scientific Acros
275020050 |
5 g | Glass bottle | N/A | ||||||
10389100
|
Thermo Scientific Acros
275022500 |
250 g | Glass bottle |
£45.53
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10287640
|
Thermo Scientific Acros
275020010 |
1 kg | Glass bottle |
£128.64
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
12054-48-7 | |
92.71 | |
BFDHFSHZJLFAMC-UHFFFAOYSA-L | |
[OH-].[OH-].[Ni++] |
H2NiO2 | |
MFCD00011140 | |
nickel(2+) dihydroxide |
Specifications
12054-48-7 | |
Crystalline Powder | |
Ni(OH)2 | |
15, 6595 | |
[OH-].[OH-].[Ni++] | |
92.71 | |
61.0 to 65.0% (Nickel) | |
Nickel(II) hydroxide, c.p., 60-61% Ni |
Green | |
H2NiO2 | |
MFCD00011140 | |
BFDHFSHZJLFAMC-UHFFFAOYSA-L | |
nickel(2+) dihydroxide | |
92.7 | |
Analytical |
Safety and Handling
GHS H Statement
May damage the unborn child. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. May cause cancer by inhalation. Harmful if inhaled. Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Use personal protective equipment as required. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
Danger
missing translation for 'einecsNumber' : 235-008-5
missing translation for 'rtecsNumber' : QR7040000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.