Learn More
Nickel(II) lactate tetrahydrate, 98%, Thermo Scientific Chemicals
£17.59 - £167.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470038
|
Thermo Scientific Alfa Aesar
B23643.14 |
25 g |
£17.59
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480038
|
Thermo Scientific Alfa Aesar
B23643.22 |
100 g |
£43.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490038
|
Thermo Scientific Alfa Aesar
B23643.36 |
500 g |
£167.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
C6H10NiO6·4H2O | |
MFCD02683106 |
Specifications
C6H10NiO6·4H2O | |
UN3077 | |
98% |
MFCD02683106 | |
308.91 (236.85 Anhydrous) | |
Nickel(II) lactate tetrahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
H317-H334-H341-H350-H360-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only