Learn More
Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Description
It is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 137°C |
Melting Point | 56.7°C |
Color | Green |
Physical Form | Crystalline Aggregates |
Quantity | 50 g |
Linear Formula | Ni(NO3)2·6H2O |
UN Number | UN2725 |
Merck Index | 14,6513 |
Solubility Information | Very soluble in water. Soluble in ethanol. |
IUPAC Name | nickel(2+);dinitrate;hexahydrate |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y11K019 | Certificate of Analysis | 19/11/2024 | 010816.06,010816.18,010816.30 |
X07K026 | Certificate of Analysis | 17/10/2024 | 010816.06,010816.18,010816.30 |
N09J024 | Certificate of Analysis | 24/10/2023 | 010816.06,010816.18,010816.30 |
U14J012 | Certificate of Analysis | 21/09/2023 | 010816.06,010816.18,010816.30 |
U15J101 | Certificate of Analysis | 06/09/2023 | 010816.06,010816.18,010816.30 |
Safety and Handling





- Nickel(II) nitrate hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- MIXTURE LIST-Contains : Nickel(II) nitrate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.