missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals Product Code.: 11365057

Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11365057
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11365057

Brand: Thermo Scientific Alfa Aesar 040757.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126956-48-7
Molecular Formula C2NiO4
Molecular Weight (g/mol) 146.71
MDL Number MFCD00167155
InChI Key DOLZKNFSRCEOFV-UHFFFAOYSA-L
Synonym nickel ii oxalate hydrate, acmc-1c3o8, nickel 2+ hydrate oxalate, nickel ii oxalate hydrate, puratronic, nickel 2+ ion hydrate oxalate, nickel 2+ ethanedioate-water 1/1/1, nickel, ethanedioato 2--ko1,ko2-, hydrate 9ciShow MoreShow Less
PubChem CID 16213068
SMILES [Ni++].[O-]C(=O)C([O-])=O

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula NiC2O4·xH2O (x≈2)
UN Number UN3077
Merck Index 14,6514
IUPAC Name nickel(2+);oxalate;hydrate
Formula Weight 146.73 (Anhydrous)
Percent Purity 99.9985%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Nickel(II) oxalate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14K051 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number040757.14,040757.06
Lot NumberN03J040 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog Number040757.14,040757.06
Lot Number94327B Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number040757.14,040757.06
Lot NumberS27F005 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040757.14,040757.06
Lot NumberW18E011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number040757.14,040757.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) oxalate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.