Learn More
Nickel(II) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Nickel (II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1984°C |
Color | Green |
Physical Form | Powder |
Quantity | 5 g |
Merck Index | 14,6512 |
Solubility Information | Soluble in ammonium hydroxide. Insoluble in water. |
Formula Weight | 74.71 |
Percent Purity | 99.998% |
Odor | Odorless |
Density | 6.67 |
Show More |
RUO – Research Use Only