Learn More
Nickel(II) sulfate, 0.5M aq. soln., Thermo Scientific Chemicals
£22.23 - £64.22
Chemical Identifiers
CAS | 10101-97-0 |
---|---|
Molecular Formula | H12NiO10S |
Molecular Weight (g/mol) | 262.839 |
MDL Number | MFCD00011146 |
InChI Key | RRIWRJBSCGCBID-UHFFFAOYSA-L |
Synonym | nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 |
PubChem CID | 5284429 |
ChEBI | CHEBI:53437 |
IUPAC Name | nickel(2+);sulfate;hexahydrate |
SMILES | O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404469
|
Thermo Scientific Alfa Aesar
J61993.AD |
50 mL | N/A | ||||||
15414469
|
Thermo Scientific Alfa Aesar
J61993.AE |
100 mL |
£64.22
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNickel(II) sulfate, 0.5M aq. soln. is used in nickel plating, as mordant in dyeing and in printing textiles. Columns used in polyhistidine-tagging, useful in biochemistry and molecular biology, are regenerated with nickel sulfate.
Solubility
Fully miscible in water.
Notes
Protect from heat.
Chemical Identifiers
10101-97-0 | |
262.839 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
5284429 | |
nickel(2+);sulfate;hexahydrate |
H12NiO10S | |
MFCD00011146 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
CHEBI:53437 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] |
Specifications
10101-97-0 | |
Liquid | |
NiSO4 | |
UN3082 | |
RRIWRJBSCGCBID-UHFFFAOYSA-L | |
nickel(2+);sulfate;hexahydrate | |
5284429 | |
262.86 | |
Odorless |
Green | |
H12NiO10S | |
MFCD00011146 | |
nickel sulfate hexahydrate, nickel ii sulfate hexahydrate, nickel 2+ sulfate hexahydrate, nickel ii sulfate, nickelsulfatehexahydrate, dsstox_cid_928, acmc-1c7xm, dsstox_rid_75872, ni.so4.6h2o, dsstox_gsid_20928 | |
O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2] | |
262.839 | |
CHEBI:53437 | |
0.5M aq. soln. | |
Nickel(II) sulfate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
P260-P284a-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'tsca' : TSCA
Recommended Storage : 2° to 8°C
RUO – Research Use Only