missing translation for 'onlineSavingsMsg'
Learn More
Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970, Thermo Scientific Chemicals Product Code.: 10267690

Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970, Thermo Scientific Chemicals

Product Code. 10267690
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10267690

Brand: Thermo Scientific Acros 270550010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-98-1, 7786-81-4
Molecular Formula H14NiO11S
Molecular Weight (g/mol) 280.85
MDL Number MFCD00149814
InChI Key OGKAGKFVPCOHQW-UHFFFAOYSA-L
Synonym nickel ii sulfate heptahydrate, nickel sulfate heptahydrate, nickel 2+ sulfate heptahydrate, nickelous sulfate heptahydrate, unii-596idd57nr, sulfuric acid, nickel 2+ salt 1:1 , heptahydrate, nickel sulfate heptahydrate nickel and nickel compounds, ni.so4.7h2o, ksc492q3n, nickel sulfate-water 1/7
PubChem CID 61474
ChEBI CHEBI:53504
SMILES O.O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

Specifications

Color Green
Physical Form Needle-like Crystals or Powder
Quantity 1 kg
Linear Formula NiSO4·7H2O
Merck Index 15, 6602
Solubility Information Solubility in water: 625g/L (20°C). Other solubilities: insoluble in alcohol and ether
IUPAC Name nickel(2+) heptahydrate sulfate
Formula Weight 280.88
Percent Purity 20 to 21% (Ni)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Nickel(II) sulfate heptahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466597 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number270550010,270552500
Lot NumberA0464281 Certificate TypeCertificate of Analysis Date05/07/2024 Catalog Number270550010,270552500
Lot NumberA0456499 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number270550010,270552500
Lot NumberA0449581 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog Number
Lot NumberA0411318 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number270550010,270552500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel(II) sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) sulfate heptahydrate, for analysis, for nickel plating, DIN 50970, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.